CH4005883320 ChemBridge Corporation

1-(3-fluorophenyl)-3-(2-hydroxy-4-methylphenyl)propane-1,3-dione

  • MDL number MFCD20502993
  • Cas Number : 332103-27-2
  • Molecular Weight : 272.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5812380

Safety Information

MSDS https://www.hit2lead.com/sds/5812380_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart