CH4005883869 ChemBridge Corporation

2-(5-bromo-2-furyl)-1H-benzimidazole

  • MDL number MFCD00453108
  • Cas Number : 18249-68-8
  • Molecular Weight : 263.0901

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5815957

Safety Information

MSDS https://www.hit2lead.com/sds/5815957_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart