CH4005887594 ChemBridge Corporation

3-(1-ethyl-1H-benzimidazol-2-yl)propan-1-ol

  • MDL number MFCD01365755
  • Cas Number : 305347-67-5
  • Molecular Weight : 204.2682

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5849051

Safety Information

MSDS https://www.hit2lead.com/sds/5849051_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart