CH4005890170 ChemBridge Corporation

1-benzoyl-2-piperidinecarboxylic acid

  • MDL number MFCD02211013
  • Cas Number : 78348-46-6
  • Molecular Weight : 233.2631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5864666

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5864666
Availability details for
Quantity
  • Availability
  • Add to Cart