CH4005891855 ChemBridge Corporation

4-(4-chloro-3-nitrophenyl)-3-methyl-4-oxobutanoic acid

  • MDL number MFCD02211210
  • Cas Number : 85633-96-1
  • Molecular Weight : 271.6538

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5876317

Safety Information

MSDS https://www.hit2lead.com/sds/5876317_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart