CH4005892979 ChemBridge Corporation

1,6-dimethyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD01918236
  • Cas Number : 51726-43-3
  • Molecular Weight : 217.2206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5884927

Safety Information

MSDS https://www.hit2lead.com/sds/5884927_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart