CH4005893371 ChemBridge Corporation

2-[5-(trifluoromethyl)-3-isoxazolyl]phenol

  • MDL number MFCD01419244
  • Cas Number : 312505-97-8
  • Molecular Weight : 229.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6001359

Safety Information

MSDS https://www.hit2lead.com/sds/6001359_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart