CH4005894971 ChemBridge Corporation

4-[(quinolin-2-ylcarbonyl)amino]benzoic acid

  • MDL number MFCD00498384
  • Cas Number : 190437-68-4
  • Molecular Weight : 292.2888

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6024454

Safety Information

MSDS https://www.hit2lead.com/sds/6024454_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart