CH4005895805 ChemBridge Corporation

2-amino-N-(2,3-dimethylphenyl)benzamide

  • MDL number MFCD01930137
  • Cas Number : 35703-71-0
  • Molecular Weight : 240.3003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6034657

Safety Information

MSDS https://www.hit2lead.com/sds/6034657_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart