CH4005897867 ChemBridge Corporation

2-(diphenylmethyl)-1,3-thiazol-4-amine

  • MDL number MFCD02108091
  • Cas Number : 425399-53-7
  • Molecular Weight : 266.3608

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6046797

Safety Information

MSDS https://www.hit2lead.com/sds/6046797_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart