CH4005903282 ChemBridge Corporation

N-(4-chlorobenzyl)-3-oxobutanamide

  • MDL number MFCD01001281
  • Cas Number : 78984-83-5
  • Molecular Weight : 225.6715

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6085090

Safety Information

MSDS https://www.hit2lead.com/sds/6085090_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart