CH4005903481 ChemBridge Corporation

dimethyl N-(4-iodobenzoyl)glutamate

  • MDL number MFCD01605810
  • Cas Number : 473444-31-4
  • Molecular Weight : 405.185

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6087257

Safety Information

MSDS https://www.hit2lead.com/sds/6087257_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart