CH4005903637 ChemBridge Corporation

1-[2-(2-methoxyphenoxy)ethyl]-1H-indole-3-carbaldehyde

  • MDL number MFCD02049624
  • Cas Number : 299936-03-1
  • Molecular Weight : 295.3325

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6088985

Safety Information

MSDS https://www.hit2lead.com/sds/6088985_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart