CH4005903798 ChemBridge Corporation

N-(2,4-difluorophenyl)-3-oxobutanamide

  • MDL number MFCD00828654
  • Cas Number : 218930-20-2
  • Molecular Weight : 213.1808

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6090969

Safety Information

MSDS https://www.hit2lead.com/sds/6090969_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart