CH4005903804 ChemBridge Corporation

N-[4-(acetylamino)phenyl]-3-oxobutanamide

  • MDL number MFCD00035810
  • Cas Number : 4433-78-7
  • Molecular Weight : 234.2512

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6091016

Safety Information

signalword Warning
hcodes 6.1
MSDS https://www.hit2lead.com/sds/6091016_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart