CH4005903834 ChemBridge Corporation

N-(2-methoxy-4-nitrophenyl)-3-oxobutanamide

  • MDL number MFCD01610158
  • Cas Number : 91088-39-0
  • Molecular Weight : 252.2234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6091432

Safety Information

MSDS https://www.hit2lead.com/sds/6091432_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart