CH4005906271 ChemBridge Corporation

N-(2-bromophenyl)-5-chloro-2-methoxybenzamide

  • MDL number MFCD01356367
  • Cas Number : 349538-99-4
  • Molecular Weight : 340.5996

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6114463

Safety Information

MSDS https://www.hit2lead.com/sds/6114463_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart