CH4005908562 ChemBridge Corporation

ethyl 2-amino-4-(4-pyridinyl)-3-thiophenecarboxylate

  • MDL number MFCD01922021
  • Cas Number : 117516-88-8
  • Molecular Weight : 248.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6132512

Safety Information

MSDS https://www.hit2lead.com/sds/6132512_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart