CH4005909738 ChemBridge Corporation

4-ethyl-5-hydroxy-7-methyl-2H-chromen-2-one

  • MDL number MFCD02080280
  • Cas Number : 62036-49-1
  • Molecular Weight : 204.2219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6141880

Safety Information

MSDS https://www.hit2lead.com/sds/6141880_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart