CH4005911615 ChemBridge Corporation

2-[(6-methyl-4-oxo-1,2,3,4-tetrahydrocyclopenta[c]chromen-7-yl)oxy]propanoic acid

  • MDL number MFCD02059336
  • Cas Number : 307548-90-9
  • Molecular Weight : 288.2952

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6157486

Safety Information

MSDS https://www.hit2lead.com/sds/6157486_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart