CH4005915958 ChemBridge Corporation

[(3,5-dimethylbenzyl)thio]acetic acid

  • MDL number MFCD02219318
  • Cas Number : 425607-97-2
  • Molecular Weight : 210.2927

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6198287

Safety Information

MSDS https://www.hit2lead.com/sds/6198287_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart