CH4005925986 ChemBridge Corporation

7-hydroxy-8-methyl-4-propyl-2H-chromen-2-one

  • MDL number MFCD02222024
  • Cas Number : 95184-99-9
  • Molecular Weight : 218.2485

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6367113

Safety Information

MSDS https://www.hit2lead.com/sds/6367113_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart