CH4005926447 ChemBridge Corporation

methyl 3-formylpyrrolo[2,1-b][1,3]benzothiazole-1-carboxylate

  • MDL number MFCD02638056
  • Cas Number : 136427-69-5
  • Molecular Weight : 259.2805

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6369811

Safety Information

MSDS https://www.hit2lead.com/sds/6369811_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart