CH4005935226 ChemBridge Corporation

ethyl 6-acetyl-2-amino-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate

  • MDL number MFCD01750487
  • Cas Number : 24237-43-2
  • Molecular Weight : 268.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6433024

Safety Information

MSDS https://www.hit2lead.com/sds/6433024_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart