CH4005936472 ChemBridge Corporation

(2E)-3-(2-furyl)-2-phenylacrylic acid

  • MDL number MFCD03135493
  • Cas Number : 42307-39-1
  • Molecular Weight : 214.2167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6442011

Safety Information

MSDS https://www.hit2lead.com/sds/6442011_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart