CH4005937369 ChemBridge Corporation

3-bromo-4-[(3-fluorobenzyl)oxy]-5-methoxybenzaldehyde

  • MDL number MFCD01990233
  • Cas Number : 346459-51-6
  • Molecular Weight : 339.1564

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6450624

Safety Information

MSDS https://www.hit2lead.com/sds/6450624_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart