CH4005937552 ChemBridge Corporation

5-nitro-2-pyrrolidin-1-ylbenzoic acid

  • MDL number MFCD01764471
  • Cas Number : 19555-48-7
  • Molecular Weight : 236.224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6452672

Safety Information

MSDS https://www.hit2lead.com/sds/6452672_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart