CH4005938469 ChemBridge Corporation

methyl 3-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzoate

  • MDL number MFCD02604825
  • Cas Number : 428473-45-4
  • Molecular Weight : 257.2845

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6463400

Safety Information

MSDS https://www.hit2lead.com/sds/6463400_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart