CH4005943152 ChemBridge Corporation

1-[3-(6-bromo-2H-1,3-benzoxazin-3(4H)-yl)phenyl]ethanone

  • MDL number MFCD02235322
  • Cas Number : 374911-94-1
  • Molecular Weight : 332.1919

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6518172

Safety Information

MSDS https://www.hit2lead.com/sds/6518172_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart