CH4005944268 ChemBridge Corporation

4-bromo-2-{[(1-propyl-1H-benzimidazol-2-yl)amino]methyl}phenol

  • MDL number MFCD02943223
  • Cas Number : 384351-72-8
  • Molecular Weight : 360.2483

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6530210

Safety Information

MSDS https://www.hit2lead.com/sds/6530210_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart