CH4005951694 ChemBridge Corporation

1-(3,4-dihydroxyphenyl)-2-[(4-hydroxy-6-methylpyrimidin-2-yl)thio]ethanone

  • MDL number MFCD02366437
  • Cas Number : 429649-34-3
  • Molecular Weight : 292.3104

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6606317

Safety Information

MSDS https://www.hit2lead.com/sds/6606317_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart