CH4005951919 ChemBridge Corporation

2-[(E)-2-(2-aminophenyl)vinyl]quinolin-8-ol

  • MDL number MFCD02923372
  • Cas Number : 429651-60-5
  • Molecular Weight : 262.3059

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6608050

Safety Information

MSDS https://www.hit2lead.com/sds/6608050_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart