CH4005953829 ChemBridge Corporation

1-(3,4-dihydroxyphenyl)-2-[(4,6-dimethylpyrimidin-2-yl)thio]ethanone

  • MDL number MFCD02366449
  • Cas Number : 430447-82-8
  • Molecular Weight : 290.3376

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6622859

Safety Information

MSDS https://www.hit2lead.com/sds/6622859_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart