CH4005956009 ChemBridge Corporation

N-(2-bromophenyl)-2-methoxyacetamide

  • MDL number MFCD03138775
  • Cas Number : 430450-95-6
  • Molecular Weight : 244.0852

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6634987

Safety Information

MSDS https://www.hit2lead.com/sds/6634987_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart