CH4005961020 ChemBridge Corporation

2-(2-chlorophenoxy)-N-(4-iodo-2-methylphenyl)acetamide

  • MDL number MFCD03140190
  • Cas Number : 431885-12-0
  • Molecular Weight : 401.6267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6672118

Safety Information

MSDS https://www.hit2lead.com/sds/6672118_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart