CH4005962323 ChemBridge Corporation

N-4-quinazolinylvaline

  • MDL number MFCD02322963
  • Cas Number : 474004-02-9
  • Molecular Weight : 245.2771

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6684905

Safety Information

MSDS https://www.hit2lead.com/sds/6684905_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart