CH4005970477 ChemBridge Corporation

N-(2-bromophenyl)-2-(4-isopropylphenoxy)acetamide

  • MDL number MFCD03143314
  • Cas Number : 431995-26-5
  • Molecular Weight : 348.2343

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6791496

Safety Information

MSDS https://www.hit2lead.com/sds/6791496_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart