CH4005970976 ChemBridge Corporation

2-bromo-6-ethoxy-4-[(E)-2-nitrovinyl]phenol

  • MDL number MFCD03143477
  • Cas Number : 432496-77-0
  • Molecular Weight : 288.0947

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6796747

Safety Information

MSDS https://www.hit2lead.com/sds/6796747_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart