CH4005972145 ChemBridge Corporation

2-bromo-6-ethoxy-4-[(1E)-2-nitroprop-1-en-1-yl]phenol

  • MDL number MFCD01461653
  • Cas Number : 312510-61-5
  • Molecular Weight : 302.1213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6809891

Safety Information

MSDS https://www.hit2lead.com/sds/6809891_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart