CH4005973483 ChemBridge Corporation

(2Z)-2-(5-bromo-2-fluorobenzylidene)[1,3]thiazolo[3,2-a]benzimidazol-3(2H)-one

  • MDL number MFCD02071609
  • Cas Number : 292169-72-3
  • Molecular Weight : 375.2149

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6827740

Safety Information

MSDS https://www.hit2lead.com/sds/6827740_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart