CH4005973875 ChemBridge Corporation

1-(2-bromophenoxy)-3-piperidin-1-ylpropan-2-ol

  • MDL number MFCD03670777
  • Cas Number : 100794-31-8
  • Molecular Weight : 314.2181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6833296

Safety Information

MSDS https://www.hit2lead.com/sds/6833296_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart