CH4005974219 ChemBridge Corporation

3-[5-(4-methylphenyl)-2-furyl]propanoic acid

  • MDL number MFCD03110272
  • Cas Number : 23589-06-2
  • Molecular Weight : 230.2592

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6838357

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6838357
Availability details for
Quantity
  • Availability
  • Add to Cart