CH4005974893 ChemBridge Corporation

4,7-dioxo-7-(2-thienyl)heptanoic acid

  • MDL number MFCD02257797
  • Cas Number : 24090-18-4
  • Molecular Weight : 240.2756

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6848429

Safety Information

MSDS https://www.hit2lead.com/sds/6848429_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart