CH4005976562 ChemBridge Corporation

4-(2,4-dimethoxyphenyl)-5-methyl-1,3-thiazol-2-amine

  • MDL number MFCD03145141
  • Cas Number : 117844-98-1
  • Molecular Weight : 250.3168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6872366

Safety Information

MSDS https://www.hit2lead.com/sds/6872366_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart