CH4005986366 ChemBridge Corporation

2-hydroxy-7-methoxyquinoline-3-carbaldehyde

  • MDL number MFCD02986395
  • Cas Number : 101382-55-2
  • Molecular Weight : 203.1941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6967832

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6967832
Availability details for
Quantity
  • Availability
  • Add to Cart