CH4005987676 ChemBridge Corporation

(2E)-3-(4-aminophenyl)-2-phenylacrylic acid

  • MDL number MFCD01414440
  • Cas Number : 52873-63-9
  • Molecular Weight : 239.2692

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6981235

Safety Information

MSDS https://www.hit2lead.com/sds/6981235_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart