CH4005989531 ChemBridge Corporation

(2-amino-4-chlorophenyl)(1-phenylethyl)amine

  • MDL number MFCD02029653
  • Cas Number : 345991-79-9
  • Molecular Weight : 246.7353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6996558

Safety Information

MSDS https://www.hit2lead.com/sds/6996558_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart