CH4005990101 ChemBridge Corporation

1-(2-chloro-4-fluorobenzyl)-1H-indole-3-carbaldehyde

  • MDL number MFCD02615000
  • Cas Number : 525570-33-6
  • Molecular Weight : 287.716

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7001766

Safety Information

MSDS https://www.hit2lead.com/sds/7001766_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart