CH4005990493 ChemBridge Corporation

4-bromo-2-(5-pyridin-3-yl-4,5-dihydro-1H-pyrazol-3-yl)phenol

  • MDL number MFCD01082642
  • Cas Number : 370074-36-5
  • Molecular Weight : 318.1686

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7003842

Safety Information

MSDS https://www.hit2lead.com/sds/7003842_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart