CH4005994411 ChemBridge Corporation

4,6-dimethyl-2-oxo-3,4-dihydro-2H-pyrido[1,2-a]pyrimidine-4-carboxylic acid

  • MDL number MFCD03449006
  • Cas Number : 956783-58-7
  • Molecular Weight : 220.2246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7043293

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7043293
Availability details for
Quantity
  • Availability
  • Add to Cart